4-(4-Methylpiperazin-1-yl)benzaldehyde

Supplier: Apollo Scientific
Warning

OR23309-1G OR23309-25G OR23309-5G
APOSOR23309-1GEA 17.1 GBP
APOSOR23309-1G APOSOR23309-25G APOSOR23309-5G
4-(4-Methylpiperazin-1-yl)benzaldehyde
4-(4-Methylpiperazin-1-yl)benzaldehyde
Formula: C₁₂H₁₆N₂O
MW: 204,2682 g/mol
Melting Pt: 41…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00452233
CAS Number: 27913-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR