6-Morpholinonicotinaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Morpholinylpyridine-3-carboxaldehyde
OR23273-1G
OR23273-250MG
APOSOR23273-1GEA
231
GBP
APOSOR23273-1G
APOSOR23273-250MG
6-Morpholinonicotinaldehyde
6-Morpholinonicotinaldehyde
Formula:
C₁₀H₁₂N₂O₂ MW: 192,22 g/mol Melting Pt: 88…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD03659703 CAS Number: 173282-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...