6-Morpholinonicotinoyl chloride, Technical Grade
Supplier: Apollo Scientific
|
Danger
|
OR23271-1G
OR23271-5G
OR23271-10G
APOSOR23271-1GEA
69.5
GBP
APOSOR23271-1G
APOSOR23271-5G
APOSOR23271-10G
6-Morpholinonicotinoyl chloride, Technical Grade
6-Morpholinonicotinoyl chloride
Formula:
C₁₀H₁₁ClN₂O₂ MW: 226,66 g/mol Melting Pt: 122…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD02681966 CAS Number: 313350-36-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...