4-Morpholinobenzoic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Morpholinobenzoic acid, 4-(4-Morpholinyl)benzoic acid
OR23266-5G
OR23266-25G
OR23266-100G
APOSOR23266-5GEA
48.6
GBP
APOSOR23266-5G
APOSOR23266-25G
APOSOR23266-100G
4-Morpholinobenzoic acid
4-Morpholinobenzoic acid
Formula:
C₁₁H₁₃NO₃ MW: 207,2300 g/mol Melting Pt: 284…289 °C Storage Temperature: Ambient |
MDL Number:
MFCD00588379 CAS Number: 7470-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...