4-Bromo-1,3,5-trimethyl-1H-pyrazole

Supplier: Apollo Scientific

Synonyms: 4-Bromo-1,3,5-trimethylpyrazole

OR23238-100G OR23238-25G
APOSOR23238-100GEA 94.4 GBP
APOSOR23238-100G APOSOR23238-25G
4-Bromo-1,3,5-trimethyl-1H-pyrazole
4-Bromo-1,3,5-trimethyl-1H-pyrazole
Formula: C₆H₉BrN₂
MW: 189,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052532
CAS Number: 15801-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR