Benzyl-4-formylpiperidine-1-carboxylate 95%

Supplier: Apollo Scientific
Warning

Synonyms: Benzyl 4-formylpiperidine-1-carboxylate, 4-Formylpiperidine, N-CBZ protected

OR23103-25G OR23103-5G
APOSOR23103-25GEA 112 GBP
APOSOR23103-25G APOSOR23103-5G
Benzyl-4-formylpiperidine-1-carboxylate 95%
Benzyl-4-formylpiperidine-1-carboxylate
Formula: C₁₄H₁₇NO₃
MW: 247,2897 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01317806
CAS Number: 138163-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR