Benzofuran-2-carbonyl chloride 97%
Supplier: Apollo Scientific
|
Danger
|
OR23094-1G
OR23094-250MG
OR23094-5G
APOSOR23094-1GEA
27.6
GBP
APOSOR23094-1G
APOSOR23094-250MG
APOSOR23094-5G
Benzofuran-2-carbonyl chloride 97%
Benzofuran-2-carbonyl chloride
Formula:
C₉H₅ClO₂ MW: 180,59 g/mol Melting Pt: 54…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD01659782 CAS Number: 41717-28-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...