3-Bromo-4-chloropyridine 97%

Supplier: Apollo Scientific
Warning

OR2306-25G OR2306-100G
APOSOR2306-25GEA 69.5 GBP
APOSOR2306-25G APOSOR2306-100G
3-Bromo-4-chloropyridine 97%
3-Bromo-4-chloropyridine
Formula: C₅H₃BrClN
Storage Temperature: Ambient
MDL Number: MFCD00234008
CAS Number: 36953-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR