2-Pyridylmethanol 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Pyridylmethanol, 2-Pyridylcarbinol, 2-Pyridinemethanol, Pyridin-2-ylmethanol, 2-(Hydroxymethyl)pyridine, Pyridine-2-methanol

OR23056-25G OR23056-100G
APOSOR23056-25GEA 14.5 GBP
APOSOR23056-25G APOSOR23056-100G
2-Pyridylmethanol 98%
2-Pyridylmethanol
Formula: C₆H₇NO
MW: 109,13 g/mol
Boiling Pt: 112…113 °C/16mmHg
Melting Pt: –6 °C
Density: 1,33 g/cm³ (25 °C)
Flash Pt: 117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006348
CAS Number: 586-98-1
EINECS: 209-592-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR