2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 1,4-Benzodioxane-6-carboxylic acid

OR23003-25G OR23003-100G
APOSOR23003-25GEA 60.3 GBP
APOSOR23003-25G APOSOR23003-100G
2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid
2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid
Formula: C₉H₈O₄
MW: 180,16 g/mol
Melting Pt: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00463509
CAS Number: 4442-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR