3-Furoyl chloride
Supplier: Apollo Scientific
|
Danger
|
OR22960-1G
OR22960-5G
APOSOR22960-1GEA
65.6
GBP
APOSOR22960-1G
APOSOR22960-5G
3-Furoyl chloride
3-Furoyl chloride
Formula:
C₅H₃ClO₂ Boiling Pt: 62…64 °C/15mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD02681970 CAS Number: 26214-65-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...