Methyl-4-aminobutanoate hydrochloride

Supplier: Apollo Scientific

Synonyms: H-γ-Abu-OMe.HCl

OR22446-100G OR22446-25G OR22446-5G
APOSOR22446-100GEA 144 GBP
APOSOR22446-100G APOSOR22446-25G APOSOR22446-5G
Methyl-4-aminobutanoate hydrochloride
Methyl-4-aminobutanoate hydrochloride
Formula: C₅H₁₁NO₂·HCl
MW: 153,61 g/mol
Melting Pt: 120…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00043270
CAS Number: 13031-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR