1-Methyl-1H-indole-3-carbaldehyde 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Methylindole-3-carboxaldehyde

OR22346-100G OR22346-25G OR22346-5G
APOSOR22346-100GEA 222 GBP
APOSOR22346-100G APOSOR22346-25G APOSOR22346-5G
1-Methyl-1H-indole-3-carbaldehyde 98%
1-Methyl-1H-indole-3-carbaldehyde
Formula: C₁₀H₉NO
MW: 159,188 g/mol
Melting Pt: 68…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014570
CAS Number: 19012-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR