4-(Ethylamino)benzoic acid

Supplier: Apollo Scientific
Warning

OR22095-10G OR22095-1G OR22095-5G
APOSOR22095-10GEA 103 GBP
APOSOR22095-10G APOSOR22095-1G APOSOR22095-5G
4-(Ethylamino)benzoic acid
4-(Ethylamino)benzoic acid
Formula: C₉H₁₁NO₂
Melting Pt: 175…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00102118
CAS Number: 7409-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR