Bis(4-hydroxyphenyl)sulphide 98%

Supplier: Apollo Scientific
Danger

Synonyms: 4,4'-Thiodiphenol

OR22056-100G OR22056-500G
APOSOR22056-100GEA 78.7 GBP
APOSOR22056-100G APOSOR22056-500G
Bis(4-hydroxyphenyl)sulphide 98%
Bis(4-hydroxyphenyl) sulphide
Formula: C₁₂H₁₀O₂S
MW: 218,28 g/mol
Melting Pt: 151…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00002349
CAS Number: 2664-63-3
EINECS: 220-197-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR