2,4-Diacetylphloroglucinol 96%

Supplier: Apollo Scientific
Warning

Synonyms: 1,1'-(2,4,6-Trihydroxy-1,3-phenylene)bisethanone, 1-(3-acetyl-2,4,6-trihydroxyphenyl)ethan-1-one, Ethanone,1,1'-(2,4,6-trihydroxy-1,3-phenylene)bis-

OR22030-1G OR22030-500MG
APOSOR22030-1GEA 643 GBP
APOSOR22030-1G APOSOR22030-500MG
2,4-Diacetylphloroglucinol 96%
2,4-Diacetylphloroglucinol
Formula: C₁₀H₁₀O₅
MW: 210,19 g/mol
Melting Pt: 164…168 °C
Storage Temperature: Freezer
MDL Number: MFCD00100483
CAS Number: 2161-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR