4-tert-Butylbenzamide 98%
Supplier: Apollo Scientific
|
Warning
|
OR21726-10G
OR21726-50G
APOSOR21726-10GEA
0
GBP
APOSOR21726-10G
APOSOR21726-50G
4-tert-Butylbenzamide 98%
4-tert-Butylbenzamide
Formula:
C₁₁H₁₅NO MW: 177,25 g/mol Melting Pt: 171…715 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017139 CAS Number: 56108-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...