2-Amino-4-methoxybenzothiazole

Supplier: Apollo Scientific
Warning

Synonyms: 4-Methoxybenzothiazol-2-ylamine

OR21286-1G OR21286-5G
APOSOR21286-1GEA 0 GBP
APOSOR21286-1G APOSOR21286-5G
2-Amino-4-methoxybenzothiazole
2-Amino-4-methoxybenzothiazole
Formula: C₈H₈N₂OS
MW: 180,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005792
CAS Number: 5464-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR