Ethyl cyclobutylideneacetate 97%

Supplier: Apollo Scientific
Warning

OR200117-1G OR200117-250MG OR200117-5G
APOSOR200117-1GEA 28.5 GBP
APOSOR200117-1G APOSOR200117-250MG APOSOR200117-5G
Ethyl cyclobutylideneacetate 97%
Ethyl cyclobutylideneacetate
Formula: C₈H₁₂O₂
Boiling Pt: 81 °C/14mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD11977311
CAS Number: 27741-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR