Dimethyl aminomalonate hydrochloride

Supplier: Apollo Scientific
Warning

Synonyms: Dimethyl 2-aminomalonate hydrochloride

OR200015-10G OR200015-1G
APOSOR200015-10GEA 0 GBP
APOSOR200015-10G APOSOR200015-1G
Dimethyl aminomalonate hydrochloride
Dimethyl aminomalonate hydrochloride
Formula: C₅H₁₀ClNO₄
MW: 183,588 g/mol
Melting Pt: 155…160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00038880
CAS Number: 16115-80-3
EINECS: 240-284-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR