1H-[1,2,3]Triazole-4-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 1H-[1,2,3]Triazole-4-carboxylic acid, 3H-[1,2,3]Triazole-4-carboxylic acid, 1H-1,2,3-Triazole-4-carboxylic acid, 3H-1-2-3-Triazole-4-carboxylic-acid, 4-Carboxy-1H-1,2,3-triazole

OR19545-5G OR19545-1G
APOSOR19545-5GEA 472 GBP
APOSOR19545-5G APOSOR19545-1G
1H-[1,2,3]Triazole-4-carboxylic acid
1H-[1,2,3]Triazole-4-carboxylic acid
Formula: C₃H₃N₃O₂
MW: 113,08 g/mol
Melting Pt: 260…261 °C
Storage Temperature: Ambient
MDL Number: MFCD00196046
CAS Number: 16681-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR