4-Bromotetrahydropyran 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromooxane, 4-Bromotetrahydro-2H-pyran

OR19531-5G OR19531-1G OR19531-25G
APOSOR19531-5GEA 48.6 GBP
APOSOR19531-5G APOSOR19531-1G APOSOR19531-25G
4-Bromotetrahydropyran 97%
4-Bromotetrahydropyran
Formula: C₅H₉BrO
MW: 165,0300 g/mol
Density: 1,467 g/cm³ (25 °C)
Flash Pt: 71,1 °C
Storage Temperature: Ambient
MDL Number: MFCD07371501
CAS Number: 25637-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR