3,5-Dichlorotoluene

Supplier: Apollo Scientific
Warning

OR19487-25G OR19487-5G OR19487-100G
APOSOR19487-25GEA 195 GBP
APOSOR19487-25G APOSOR19487-5G APOSOR19487-100G
3,5-Dichlorotoluene
3,5-Dichlorotoluene
Formula: C₇H₆Cl₂
MW: 161,03 g/mol
Boiling Pt: 78…79 °C/9mmHg
Melting Pt: 24…26 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00035819
CAS Number: 25186-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR