3-Chloro-4-iodotoluene 98%

Supplier: Apollo Scientific
Warning

OR18412-100G OR18412-25G
APOSOR18412-100GEA 210 GBP
APOSOR18412-100G APOSOR18412-25G
3-Chloro-4-iodotoluene 98%
3-Chloro-4-iodotoluene
Formula: C₇H₆ClI
MW: 252,48 g/mol
Boiling Pt: 140 °C/22mmHg
Density: 1,82 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00079734
CAS Number: 116632-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR