Cytosine 98%
Supplier: Apollo Scientific
Synonyms:
6-Aminopyrimidin-2(1H)-one, 4-Amino-2-pyrimidinol
OR18333-100G
OR18333-1KG
OR18333-500G
APOSOR18333-100GEA
15.8
GBP
APOSOR18333-100G
APOSOR18333-1KG
APOSOR18333-500G
Cytosine 98%
Cytosine
Formula:
C₄H₅N₃O MW: 111,1 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006034 CAS Number: 71-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...