1-Ethyl-4-piperidone 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Ethylpiperidin-4-one

OR18292-25G OR18292-100G
APOSOR18292-25GEA 85.1 GBP
APOSOR18292-25G APOSOR18292-100G
1-Ethyl-4-piperidone 98%
1-Ethyl-4-piperidone
Formula: C₇H₁₃NO
Boiling Pt: 46…48 °C/1mmHg
Density: 0,944 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00013356
CAS Number: 3612-18-8
EINECS: 222-781-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR