3-Aminopyrazine-2-carboxylic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Amino-2-pyrazinecarboxylic acid
OR18277-100G
OR18277-25G
OR18277-500G
APOSOR18277-100GEA
76.1
GBP
APOSOR18277-100G
APOSOR18277-25G
APOSOR18277-500G
3-Aminopyrazine-2-carboxylic acid 98%
3-Aminopyrazine-2-carboxylic acid
Formula:
C₅H₅N₃O₂ MW: 139,11 g/mol Melting Pt: 205…210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006141 CAS Number: 5424-01-1 EINECS: 226-558-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...