5-Chloro-2-benzoxazolinone 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Chloro-2(3H)-benzoxazolone, 5-Chlorobenzo[d]oxazol-2(3H)-one, Chlorzoxazone, Chloroxazone
OR18273-100G
OR18273-250G
APOSOR18273-100GEA
62.9
GBP
APOSOR18273-100G
APOSOR18273-250G
5-Chloro-2-benzoxazolinone 99%
5-Chloro-2-benzoxazolinone
Formula:
C₇H₄ClNO₂ MW: 169,57 g/mol Melting Pt: 191…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005717 CAS Number: 95-25-0 EINECS: 202-403-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...