Methyl-5-aminonicotinate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 5-aminonicotinate, Methyl 5-aminopyridine-3-carboxylate

OR18117-10G OR18117-25G
APOSOR18117-10GEA 94.4 GBP
APOSOR18117-10G APOSOR18117-25G
Methyl-5-aminonicotinate
Methyl-5-aminonicotinate
Formula: C₇H₈N₂O₂
MW: 152,1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038688
CAS Number: 36052-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR