5-Bromo-2-methyl-3-nitroaniline 95+%

Supplier: Apollo Scientific
Warning

OR17615-10G OR17615-5G OR17615-25G
APOSOR17615-10GEA 284 GBP
APOSOR17615-10G APOSOR17615-5G APOSOR17615-25G
5-Bromo-2-methyl-3-nitroaniline 95+%
5-Bromo-2-methyl-3-nitroaniline
Formula: C₇H₇BrN₂O₂
MW: 231,0500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781362
CAS Number: 864550-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR