1-(5-Nitropyridin-2-yl)piperazine 95%

Supplier: Apollo Scientific
Warning

OR1756-500MG
APOSOR1756-500MGEA 19.7 GBP
APOSOR1756-500MG
1-(5-Nitropyridin-2-yl)piperazine 95%
1-(5-Nitropyridin-2-yl)piperazine
Formula: C₉H₁₂N₄O₂
MW: 208.2200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053059
CAS Number: 82205-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR