6-Bromo-2-methoxyquinoline 96%
Supplier: Apollo Scientific
|
Warning
|
OR17559-1G
OR17559-250MG
APOSOR17559-1GEA
108
GBP
APOSOR17559-1G
APOSOR17559-250MG
6-Bromo-2-methoxyquinoline 96%
6-Bromo-2-methoxyquinoline
Formula:
C₁₀H₈BrNO MW: 238,08 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11847805 CAS Number: 99455-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...