1-Methyl-1H-pyrazole-5-boronic acid pinacol ester

Supplier: Apollo Scientific
Warning

Synonyms: 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 4,4,5,5-Tetramethyl-2-(1-methylpyrazol-5-yl)-1,3,2-dioxaborolane, 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Methylpyrazole-5-boronic acid Pinacol ester

OR17209-1G OR17209-5G
APOSOR17209-1GEA 19.7 GBP
APOSOR17209-1G APOSOR17209-5G
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester
Formula: C₁₀H₁₇BN₂O₂
MW: 208,07 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD05861380
CAS Number: 847818-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR