4-Bromo-3,5-dimethylbenzonitrile 98%

Supplier: Apollo Scientific
Warning

OR17111-10G OR17111-25G OR17111-5G
APOSOR17111-10GEA 104 GBP
APOSOR17111-10G APOSOR17111-25G APOSOR17111-5G
4-Bromo-3,5-dimethylbenzonitrile 98%
4-Bromo-3,5-dimethylbenzonitrile
Formula: C₉H₈BrN
MW: 210,0700 g/mol
Melting Pt: 144…145 °C
Storage Temperature: Ambient
MDL Number: MFCD07369910
CAS Number: 75344-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR