3-Amino-4-nitrobenzoic acid

Supplier: Apollo Scientific

OR17019-1G OR17019-5G
APOSOR17019-1GEA 106 GBP
APOSOR17019-1G APOSOR17019-5G
3-Amino-4-nitrobenzoic acid
3-Amino-4-nitrobenzoic acid
Formula: C₇H₆N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD01806270
CAS Number: 6968-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR