6-Bromo-2-oxo-1,2-dihydro-3-quinolinecarbonitrile

Supplier: Apollo Scientific

Synonyms: 6-Bromo-1,2-dihydro-2-oxoquinoline-3-carbonitrile

OR16899-1G OR16899-500MG
APOSOR16899-1GEA 348 GBP
APOSOR16899-1G APOSOR16899-500MG
6-Bromo-2-oxo-1,2-dihydro-3-quinolinecarbonitrile
6-Bromo-2-oxo-1,2-dihydro-3-quinolinecarbonitrile
Formula: C₁₀H₅BrN₂O
MW: 249,07 g/mol
Melting Pt: 308…311 °C
Storage Temperature: Ambient
MDL Number: MFCD09724162
CAS Number: 99465-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR