2-Amino-5-bromobenzaldehyde

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromoanthranilaldehyde

OR16893-1G OR16893-5G
APOSOR16893-1GEA 69.5 GBP
APOSOR16893-1G APOSOR16893-5G
2-Amino-5-bromobenzaldehyde
2-Amino-5-bromobenzaldehyde
Formula: C₇H₆BrNO
MW: 200,00 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD10696879
CAS Number: 29124-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR