7-Methylindole-3-carboxyaldehyde 98%

Supplier: Apollo Scientific

Synonyms: 7-Methyl-1H-indole-3-carbaldehyde

OR1668-1G OR1668-5G
APOSOR1668-1GEA 51.2 GBP
APOSOR1668-1G APOSOR1668-5G
7-Methylindole-3-carboxyaldehyde 98%
7-Methylindole-3-carboxyaldehyde
Formula: C₁₀H₉NO
MW: 159,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047170
CAS Number: 4771-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR