2-Bromo-5-methoxybenzonitrile

Supplier: Apollo Scientific
Warning

OR16481-1G OR16481-5G OR16481-10G
APOSOR16481-1GEA 85.2 GBP
APOSOR16481-1G APOSOR16481-5G APOSOR16481-10G
2-Bromo-5-methoxybenzonitrile
2-Bromo-5-methoxybenzonitrile
Formula: C₈H₆BrNO
MW: 212,05 g/mol
Melting Pt: 99…102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861260
CAS Number: 138642-47-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR