Benzenesulphonic acid 94%

Supplier: Apollo Scientific
Danger

OR16405-2.5KG OR16405-500G
APOSOR16405-2.5KGEA 199 GBP
APOSOR16405-2.5KG APOSOR16405-500G
Benzenesulphonic acid 94%
Benzenesulphonic acid
Formula: C₆H₆O₃S
MW: 158,18 g/mol
Melting Pt: 43…44 °C
Density: 1,32 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00011689
CAS Number: 98-11-3
EINECS: 202-638-7
UN: 2585
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR