2-Pyridineboronic acid pinacol ester 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-pyridine
OR15598-1G
OR15598-500MG
APOSOR15598-1GEA
176
GBP
APOSOR15598-1G
APOSOR15598-500MG
2-Pyridineboronic acid pinacol ester 96%
2-Pyridineboronic acid pinacol ester
Formula:
C₁₁H₁₆BNO₂ MW: 205,06 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD04039342 CAS Number: 874186-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...