5-(Chloromethyl)-4-methyl-1H-imidazole hydrochloride
Supplier: Apollo Scientific
|
Warning
|
OR15570-1G
OR15570-250MG
APOSOR15570-1GEA
217
GBP
APOSOR15570-1G
APOSOR15570-250MG
5-(Chloromethyl)-4-methyl-1H-imidazole hydrochloride
5-(Chloromethyl)-4-methyl-1H-imidazole hydrochloride
Formula:
C₅H₈Cl₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD00159660 CAS Number: 51605-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...