4-(Chloromethyl)thiazole hydrochloride

Supplier: Apollo Scientific
Danger

OR15568-100G OR15568-25G OR15568-5G
APOSOR15568-100GEA 439 GBP
APOSOR15568-100G APOSOR15568-25G APOSOR15568-5G
4-(Chloromethyl)thiazole hydrochloride
4-(Chloromethyl)thiazole hydrochloride
Formula: C₄H₄ClNS
MW: 170.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094738
CAS Number: 7709-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR