4-Bromo-2-methoxythiazole

Supplier: Apollo Scientific
Warning

OR15560-1G OR15560-5G
APOSOR15560-1GEA 72 GBP
APOSOR15560-1G APOSOR15560-5G
4-Bromo-2-methoxythiazole
4-Bromo-2-methoxythiazole
Formula: C₄H₄BrNOS
MW: 194.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07787374
CAS Number: 240816-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR