5-Bromo-2-ethoxypyrimidine
Supplier: Apollo Scientific
|
Warning
|
OR15514-1G
OR15514-5G
APOSOR15514-1GEA
110
GBP
APOSOR15514-1G
APOSOR15514-5G
5-Bromo-2-ethoxypyrimidine
5-Bromo-2-ethoxypyrimidine
Formula:
C₆H₇BrN₂O MW: 203.0400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07375135 CAS Number: 17758-11-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...