5-Chloro-2-methylthiophenol
Supplier: Apollo Scientific
OR15483-5G
OR15483-25G
APOSOR15483-5GEA
129
GBP
APOSOR15483-5G
APOSOR15483-25G
5-Chloro-2-methylthiophenol
5-Chloro-2-methylthiophenol
Formula:
C₇H₇ClS Storage Temperature: Ambient |
MDL Number:
MFCD08062429 CAS Number: 18858-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...