4,6,7,8-Tetrahydro-2,5(1H,3H)-quinolinedione

Supplier: Apollo Scientific

Synonyms: 3,4,7,8-Tetrahydroquinoline-2,5(1H,6H)-dione

OR15364-1G OR15364-5G
APOSOR15364-1GEA 55.1 GBP
APOSOR15364-1G APOSOR15364-5G
4,6,7,8-Tetrahydro-2,5(1H,3H)-quinolinedione
4,6,7,8-Tetrahydro-2,5(1H,3H)-quinolinedione
Formula: C₉H₁₁NO₂
MW: 165,19 g/mol
Melting Pt: 196…198 °C
Storage Temperature: Ambient
MDL Number: MFCD01111799
CAS Number: 5057-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR