2-Methyl-1,3-benzoxazol-4-amine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Amino-2-methyl-1,3-benzoxazole
OR15351-1G
OR15351-5G
APOSOR15351-1GEA
138
GBP
APOSOR15351-1G
APOSOR15351-5G
2-Methyl-1,3-benzoxazol-4-amine
2-Methyl-1,3-benzoxazol-4-amine
Formula:
C₁₁H₁₄N₂O Melting Pt: 76…78 °C Storage Temperature: Ambient |
CAS Number:
342897-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...