4-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid

Supplier: Apollo Scientific

OR15316-1G OR15316-5G
APOSOR15316-1GEA 112 GBP
APOSOR15316-1G APOSOR15316-5G
4-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid
4-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid
Formula: C₆H₆BrNO₂
MW: 204,0213 g/mol
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD07838483
CAS Number: 875160-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR