4-(tert-Butylsulphanyl)benzenecarbaldehyde

Supplier: Apollo Scientific
Warning

OR15248-1G OR15248-5G
APOSOR15248-1GEA 165 GBP
APOSOR15248-1G APOSOR15248-5G
4-(tert-Butylsulphanyl)benzenecarbaldehyde
4-(tert-Butylsulphanyl)benzenecarbaldehyde
Formula: C₁₁H₁₄OS
MW: 194,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443944
CAS Number: 88357-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR